Triisobutylene (mixture of branched chain isomer) ≥90.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T0456-25ML T0456-500ML
TCIAT0456-25MLEA 17.6 EUR
TCIAT0456-25ML TCIAT0456-500ML
Triisobutylene (mixture of branched chain isomer) ≥90.0% (by GC)
Triisobutylene (mixture of branched chain isomer)
Formule: C₁₂H₂₄
Kookpunt: 177°C
Vlampunt: 50°C
MDL Nummer: MFCD00060236
CAS nummer: 7756-94-7
EINECS: 500-001-0
UN: 2324
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR