Triphenylphosphine ≥95.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T0519-25G T0519-500G T0519-100G
TCIAT0519-25GEA 18.7 EUR
TCIAT0519-25G TCIAT0519-500G TCIAT0519-100G
Triphenylphosphine ≥95.0% (by titrimetric analysis)
Triphenylphosphine
Formule: C₁₈H₁₅P
Kookpunt: 260°C
Smeltpunt: 79 °C
Dichtheid: 1,194 g/cm³ (20 °C)
Vlampunt: 180 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003043
CAS nummer: 603-35-0
EINECS: 210-036-0
Merck Index: 14,09743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebe0c3e17ddb3c6f12093e1915d0d56d67894eb&vid=aebe0c3e17ddb3c6f12093e1915d0d56d67894eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Melting point 80.0 to 82.0 deg-C
Solubility in Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR