L(-)-Tryptophan ≥98.5% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T0541-25G T0541-100G
TCIAT0541-25GEA 30.8 EUR
TCIAT0541-25G TCIAT0541-100G
L(-)-Tryptophan ≥98.5% (by HPLC, titration analysis)
L(-)-Tryptophan
Formule: C₁₁H₁₂N₂O₂
Smeltpunt: 282°C
Dichtheid: 1,34 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064340
CAS nummer: 73-22-3
EINECS: 200-795-6
Merck Index: 14,09797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f44e30d00cde36d47fc85a8a8129982519fc6b42&vid=d978e6cf9017173208e6b5d7d78d76c69f5df6e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.5 area%
Purity (Nonaqueous Titration) min. 98.5 %
Specific rotation [a]20/D -30.5 to -32.5 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR