N,N,N',N'-Tetramethyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T0548-25ML T0548-500ML T0548-100ML
TCIAT0548-25MLEA 17.6 EUR
TCIAT0548-25ML TCIAT0548-500ML TCIAT0548-100ML
N,N,N',N'-Tetramethyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
1,3-Bis(dimethylamino)propane
Formule: C₇H₁₈N₂
Kookpunt: 145°C
Dichtheid: 0,78 g/cm³ (20 °C)
Vlampunt: 56°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008337
CAS nummer: 110-95-2
EINECS: 203-818-8
UN: 2734
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea10b11efe00e77c0b6a9dcd0845f5d0e557a622&vid=ea10b11efe00e77c0b6a9dcd0845f5d0e557a622&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR