L(-)-Tyrosine ≥98.5% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T0550-25G T0550-500G T0550-100G
TCIAT0550-25GEA 37.4 EUR
TCIAT0550-25G TCIAT0550-500G TCIAT0550-100G
L(-)-Tyrosine ≥98.5% (by titrimetric analysis)
L(-)-Tyrosine
Formule: C₉H₁₁NO₃
Kookpunt: 351 °C (1013 hPa)
Smeltpunt: 285 °C
Dichtheid: 1,41 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002606
CAS nummer: 60-18-4
EINECS: 200-460-4
Merck Index: 14,09839
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a875abe54f89cdad0d5a6d4de40d61c6ced0c3d3&vid=a875abe54f89cdad0d5a6d4de40d61c6ced0c3d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.5 %
Solubility in 1mol/L HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR