Triphenylphosphine oxide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T0625-25G T0625-500G T0625-100G
TCIAT0625-25GEA 18.7 EUR
TCIAT0625-25G TCIAT0625-500G TCIAT0625-100G
Triphenylphosphine oxide ≥98.0% (by GC)
Triphenylphosphine oxide
Formule: C₁₈H₁₅OP
MW: 278.29 g/mol
Vlampunt: 180
MDL Nummer: MFCD00002080
CAS nummer: 791-28-6
EINECS: 212-338-8
UN: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=270c01dafb4113daf9a0693feaf896703c3d83fd&vid=270c01dafb4113daf9a0693feaf896703c3d83fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 155.0 to 159.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR