Tetrabutylthiuram disulphide ≥98.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T0635-25G T0635-500G
TCIAT0635-25GEA 17.6 EUR
TCIAT0635-25G TCIAT0635-500G
Tetrabutylthiuram disulphide ≥98.0% (by total nitrogen basis)
Tetrabutylthiuram disulphide
Formule: C₁₈H₃₆N₂S₄
MDL Nummer: MFCD00027191
CAS nummer: 1634-02-2
EINECS: 216-652-6

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR