Tricine ≥99.0% (by titrimetric analysis) Good's buffer component for biological research

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T0682-25G T0682-250G
TCIAT0682-25GEA 28.6 EUR
TCIAT0682-25G TCIAT0682-250G
Tricine ≥99.0% (by titrimetric analysis) Good's buffer component for biological research
Tricine
Formule: (HOCH₂)₃CNHCH₂CO₂H
Smeltpunt: 187 °C
Dichtheid: 1,523 g/cm³ (19.85 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004277
CAS nummer: 5704-04-1
EINECS: 227-193-6
Merck Index: 14,09651
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66a1bbba6c838b918da6a8aca9f5c25fb4a73a66&vid=63c4c67704ef3a1a4ab1a87e299da20d212eaecb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Absorbance max. 0.30 (0.1mol/L、H2O、260nm)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR