Taurocholic acid sodium salt (from bovine bile) ≥84% (by NMR)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T0808-1G T0808-25G T0808-5G
TCIAT0808-1GEA 40.7 EUR
TCIAT0808-1G TCIAT0808-25G TCIAT0808-5G
Taurocholic acid sodium salt (from bovine bile) ≥84% (by NMR)
Taurocholic acid sodium salt
Formule: C₂₆H₄₄NNaO₇S
MDL Nummer: MFCD00013050
CAS nummer: 145-42-6
EINECS: 205-653-7
Merck Index: 14,09075

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (qNMR) min. 84.0 %
Specific rotation [a]20/D +20.0 to +26.0 deg (C=3, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR