Tetrabutylammonium perchlorate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T0836-25G T0836-500G T0836-100G
TCIAT0836-25GEA 51.7 EUR
TCIAT0836-25G TCIAT0836-500G TCIAT0836-100G
Tetrabutylammonium perchlorate ≥98.0% (by titrimetric analysis)
Tetrabutylammonium perchlorate
Formule: C₁₆H₃₆ClNO₄
Smeltpunt: 210 °C
Dichtheid: 1,07 g/cm³ (20 °C)
MDL Nummer: MFCD00038722
CAS nummer: 1923-70-2
EINECS: 217-655-5
UN: 1479
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=297a6a3b04a7287f82b5518dce8db8a79437fe51&vid=297a6a3b04a7287f82b5518dce8db8a79437fe51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (Ion exchange titration) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR