Tetraethylammonium tetrahydroborate ≥90.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Tetraethylammonium borohydride, Tetraethylammonium tetrahydridoborate

T0837-25G T0837-5G
TCIAT0837-25GEA 200 EUR
TCIAT0837-25G TCIAT0837-5G
Tetraethylammonium tetrahydroborate ≥90.0% (by titrimetric analysis)
Tetraethylammonium tetrahydroborate
Formule: C₈H₂₄BN
MW: 145.1 g/mol
MDL Nummer: MFCD00031600
CAS nummer: 17083-85-1
EINECS: 241-140-4
UN: 3131
ADR: 4.3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=674019dff4dddd420623dbb68c836cb849fe930b&vid=674019dff4dddd420623dbb68c836cb849fe930b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Sodium hypochlorite Method) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR