Tetrapropylammonium hydroxide 20 - 25% in water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T1020-25ML T1020-500ML T1020-100ML
TCIAT1020-25MLEA 27.5 EUR
TCIAT1020-25ML TCIAT1020-500ML TCIAT1020-100ML
Tetrapropylammonium hydroxide 20 - 25% in water
Tetrapropylammonium hydroxide
Formule: C₁₂H₂₉NO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009360
CAS nummer: 4499-86-9
EINECS: 224-800-6
UN: 3267
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=453b68e0acc5ce0e39edc269272904ffe0944ec6&vid=31d9e72df41ea6203fb817e211de8f94161d6708&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Concentration (Neutralization titration) 20.0 to 25.0 w/w%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR