Trichloroacetyl isocyanate ≥97.0% (by total nitrogen basis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T1047-1G T1047-25G T1047-5G
TCIAT1047-1GEA 26.4 EUR
TCIAT1047-1G TCIAT1047-25G TCIAT1047-5G
Trichloroacetyl isocyanate ≥97.0% (by total nitrogen basis)
Trichloroacetyl isocyanate
Formule: C₃Cl₃NO₂
Kookpunt: 80 - 85 °C (27 hPa)
Dichtheid: 1,59 g/cm³ (20 °C)
Vlampunt: 65°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002033
CAS nummer: 3019-71-4
EINECS: 221-165-7
UN: 2206
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1bfa9a63b4d80b011e1c3452acb1e9d642d93bb&vid=e1bfa9a63b4d80b011e1c3452acb1e9d642d93bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (with Total Nitrogen) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR