4,4',4''-Tris(benzoyloxy)trityl bromide ≥97.0% (by titrimetric analysis) hydroxyl protecting agent

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T1071-25G T1071-5G
TCIAT1071-25GEA 1330 EUR
TCIAT1071-25G TCIAT1071-5G
4,4',4''-Tris(benzoyloxy)trityl bromide ≥97.0% (by titrimetric analysis) hydroxyl protecting agent
4,4',4''-Tris(benzoyloxy)trityl bromide
Formule: C₄₀H₂₇BrO₆
MDL Nummer: MFCD00010302
CAS nummer: 86610-66-4

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (Argentmetric Titration) min. 97.0 %
Melting point 192.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR