2',4',6'-Trimethylacetophenone ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T1117-25ML T1117-5ML
TCIAT1117-25MLEA 68.2 EUR
TCIAT1117-25ML TCIAT1117-5ML
2',4',6'-Trimethylacetophenone ≥97.0% (by GC)
2',4',6'-Trimethylacetophenone
Formule: C₁₁H₁₄O
Kookpunt: 236°C
Dichtheid: 0,975 g/cm³ (20 °C)
MDL Nummer: MFCD00008735
CAS nummer: 1667-01-2
EINECS: 216-783-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebbf8cf43ca50523ae7348c30ea646f54130d9f1&vid=ebbf8cf43ca50523ae7348c30ea646f54130d9f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR