α,α,α-Trifluoro-p-toluic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T1145-25G T1145-5G
TCIAT1145-25GEA 118 EUR
TCIAT1145-25G TCIAT1145-5G
α,α,α-Trifluoro-p-toluic acid ≥98.0% (by titrimetric analysis)
α,α,α-Trifluoro-p-toluic acid
Formule: C₈H₅F₃O₂
Smeltpunt: 219…220 °C
MDL Nummer: MFCD00002562
CAS nummer: 455-24-3
EINECS: 207-242-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ab985f21224bb5b354c8896c920150184565a8e&vid=3ab985f21224bb5b354c8896c920150184565a8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 221.0 to 225.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR