Tetrabutylammonium salicylate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Tetrabutylammonium 2-hydroxybenzoate

T1276-25G
TCIAT1276-25GEA 82.5 EUR
TCIAT1276-25G
Tetrabutylammonium salicylate ≥98.0% (by HPLC)
Tetrabutylammonium salicylate
Formule: C₂₃H₄₁NO₃
MW: 379.59 g/mol
MDL Nummer: MFCD00145388
CAS nummer: 22307-72-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR