D-(-)-Diisopropyl tartrate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: (2S,3S)-(-)-Diisopropyl tartrate, (2S,3S)-Diisopropyl 2,3-dihydroxysuccinate, D-(-)-Tartaric acid diisopropyl ester

T1387-25G T1387-250G
TCIAT1387-25GEA 77 EUR
TCIAT1387-25G TCIAT1387-250G
D-(-)-Diisopropyl tartrate ≥98.0% (by GC)
D-(-)-Diisopropyl tartrate
Formule: C₁₀H₁₈O₆
MW: 234.25 g/mol
MDL Nummer: MFCD00008876
CAS nummer: 62961-64-2
EINECS: 263-771-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ba45e15a30fd461e796daaca6cf5cc6726274e2&vid=3ba45e15a30fd461e796daaca6cf5cc6726274e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -14.0 to -19.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR