o-Tolualdehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2'-Methylbenzaldehyde 2,4-Dinitrophenylhydrazone

T1447-1G T1447-10G
TCIAT1447-1GEA 79.2 EUR
TCIAT1447-1G TCIAT1447-10G
o-Tolualdehyde 2,4-Dinitrophenylhydrazone ≥98.0% (by HPLC, titration analysis)
o-Tolualdehyde 2,4-Dinitrophenylhydrazone
Formule: C₁₄H₁₂N₄O₄
MW: 300.27 g/mol
MDL Nummer: MFCD00191584
CAS nummer: 1773-44-0
UN: 1325
ADR: 4.1 III

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 195.0 to 198.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR