Tetraphenylphosphonium iodide ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T1450-10G
TCIAT1450-10GEA 93.5 EUR
TCIAT1450-10G
Tetraphenylphosphonium iodide ≥98.0% (by HPLC)
Tetraphenylphosphonium iodide
Formule: C₂₄H₂₀IP
Smeltpunt: 338…340 °C
Dichtheid: 1,5 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011917
CAS nummer: 2065-67-0
EINECS: 218-179-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=820d9736b5c5b37c6c09c96808acf7fd47740a6b&vid=820d9736b5c5b37c6c09c96808acf7fd47740a6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR