Octadecyl-p-toluenesulphonate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T1471-5G
TCIAT1471-5GEA 135 EUR
TCIAT1471-5G
Octadecyl-p-toluenesulphonate ≥98.0% (by GC)
Octadecyl-p-toluenesulphonate
Formule: C₂₅H₄₄O₃S
MDL Nummer: MFCD00191588
CAS nummer: 3386-32-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR