5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin ≥95.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T1497-100MG
TCIAT1497-100MGEA 68.2 EUR
TCIAT1497-100MG
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin ≥95.0% (by HPLC)
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin
Formule: C₄₄H₃₀N₄O₄
MDL Nummer: MFCD00191501
CAS nummer: 51094-17-8

Order Now

Specification Test Results

Appearance Gray to Dark purple to Black powder to crystal
Purity (HPLC) min. 95.0 area%
Lambda max. 416.0 to 420.0 nm (MeOH)
Molar extinction coefficient min. 400000 (MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR