Tributylsulphonium iodide ≥96.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T1564-1G
TCIAT1564-1GEA 104 EUR
TCIAT1564-1G
Tributylsulphonium iodide ≥96.0% (by titrimetric analysis)
Tributylsulphonium iodide
Formule: C₁₂H₂₇IS
MDL Nummer: MFCD00671571
CAS nummer: 18146-62-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Periodic acid Method) min. 96.0 %
Melting point 93.0 to 95.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR