1,1,2,2-Tetrachloro-1,2-dimethyldisilane ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T1590-25G T1590-5G
TCIAT1590-25GEA 434 EUR
TCIAT1590-25G TCIAT1590-5G
1,1,2,2-Tetrachloro-1,2-dimethyldisilane ≥98.0% (by titrimetric analysis)
1,1,2,2-Tetrachloro-1,2-dimethyldisilane
Formule: C₂H₆Cl₄Si₂
Vlampunt: 49°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00192469
CAS nummer: 4518-98-3
EINECS: 224-844-6
UN: 2988
ADR: 4.3 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba7b953a1101de2e887729e216cd053d5bc570ac&vid=ba7b953a1101de2e887729e216cd053d5bc570ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR