Tri(2-furyl)phosphine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T1643-1G T1643-5G
TCIAT1643-1GEA 50.6 EUR
TCIAT1643-1G TCIAT1643-5G
Tri(2-furyl)phosphine ≥98.0% (by GC)
Tri(2-furyl)phosphine
Formule: C₁₂H₉O₃P
Kookpunt: 136°C
MDL Nummer: MFCD00151857
CAS nummer: 5518-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a59699b7d0a1269f1ea8739848a3e0d6422bd900&vid=a59699b7d0a1269f1ea8739848a3e0d6422bd900&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR