TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T1656-1G T1656-25G T1656-5G
TCIAT1656-1GEA 27.5 EUR
TCIAT1656-1G TCIAT1656-25G TCIAT1656-5G
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) ≥98.0% (by titrimetric analysis)
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)
Formule: C₉H₁₆ClO₆P
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00145469
CAS nummer: 51805-45-9
ADR: 8,II
Merck Index: 14,09085
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd5ee29f8f3e3f653187de712be2099096c31ce5&vid=cd5ee29f8f3e3f653187de712be2099096c31ce5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR