Tri(2-thienyl)phosphine ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T1666-1G T1666-5G
TCIAT1666-1GEA 92.4 EUR
TCIAT1666-1G TCIAT1666-5G
Tri(2-thienyl)phosphine ≥96.0% (by GC)
Tri(2-thienyl)phosphine
Formule: C₁₂H₉PS₃
Kookpunt: 205°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00015471
CAS nummer: 24171-89-9
EINECS: 246-059-8

Order Now

Specification Test Results

Appearance White to Amber to Dark green powder to lump
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR