1-(Triisopropylsilyl)pyrrole ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T1677-25ML T1677-5ML
TCIAT1677-25MLEA 294 EUR
TCIAT1677-25ML TCIAT1677-5ML
1-(Triisopropylsilyl)pyrrole ≥96.0% (by GC)
1-(Triisopropylsilyl)pyrrole
Formule: C₁₃H₂₅NSi
Vlampunt: 107°C
MDL Nummer: MFCD00054932
CAS nummer: 87630-35-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR