2-Hydroxyethyl methyl terephthalate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Terephthalic Acid 2-Hydroxyethyl Methyl Ester

T1679-5G T1679-1G
TCIAT1679-5GEA 297 EUR
TCIAT1679-5G TCIAT1679-1G
2-Hydroxyethyl methyl terephthalate ≥97.0% (by GC)
2-Hydroxyethyl methyl terephthalate
Formule: C₁₁H₁₂O₅
MW: 224.21 g/mol
MDL Nummer: MFCD01321191
CAS nummer: 3645-00-9
EINECS: 222-877-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 81.0 to 84.0 deg-C
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR