Tetrabutylammonium hydroxide 40% in water

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T1685-500G T1685-25G T1685-100G
TCIAT1685-500GEA 286 EUR
TCIAT1685-500G TCIAT1685-25G TCIAT1685-100G
Tetrabutylammonium hydroxide 40% in water
Tetrabutylammonium hydroxide
Formule: C₁₆H₃₇NO
MW: 259.48 g/mol
Smeltpunt: 29.85
Dichtheid: 0.968
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009425
CAS nummer: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0646a071b38d05126ef6f72baf09baa5245cf1a9&vid=f1e3ae31bc95cd81b3753242e838c18c4bf34687&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Concentration (Neutralization titration) 39.5 to 44.5 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR