N-(1,1,3,3-Tetramethylbutyl)formamide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T1758-5G
TCIAT1758-5GEA 71.5 EUR
TCIAT1758-5G
N-(1,1,3,3-Tetramethylbutyl)formamide ≥98.0% (by GC)
N-(1,1,3,3-Tetramethylbutyl)formamide
Formule: C₉H₁₉NO
Kookpunt: 110°C
MDL Nummer: MFCD00039569
CAS nummer: 10151-02-7
EINECS: 233-419-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR