(α,α,α-Trifluoro-4-tolyl)acetic acid ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T1783-1G T1783-25G T1783-5G
TCIAT1783-1GEA 25.3 EUR
TCIAT1783-1G TCIAT1783-25G TCIAT1783-5G
(α,α,α-Trifluoro-4-tolyl)acetic acid ≥97.0% (by titrimetric analysis)
(α,α,α-Trifluoro-4-tolyl)acetic acid
Formule: (C₆H₄(CF₃))CH₂COOH
MDL Nummer: MFCD00004352
CAS nummer: 32857-62-8
EINECS: 251-263-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6799ddd765529c97c88ae374f510af358a291c61&vid=6799ddd765529c97c88ae374f510af358a291c61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 82.0 to 85.0 deg-C
Solubility in Methanol very faint turbidity

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR