2,4,6-Trimethylbenzyl alcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T1905-25G T1905-5G
TCIAT1905-25GEA 312 EUR
TCIAT1905-25G TCIAT1905-5G
2,4,6-Trimethylbenzyl alcohol ≥98.0% (by GC)
2,4,6-Trimethylbenzyl alcohol
Formule: C₁₀H₁₄O
Kookpunt: 141°C
Smeltpunt: 88 °C
MDL Nummer: MFCD00014422
CAS nummer: 4170-90-5
EINECS: 224-032-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=412b933f0a22620cac643469faf0f4c1715266b4&vid=412b933f0a22620cac643469faf0f4c1715266b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 90.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR