2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T1943-1G T1943-5G
TCIAT1943-1GEA 126 EUR
TCIAT1943-1G TCIAT1943-5G
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid ≥98.0% (by titrimetric analysis)
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid
Formule: C₇H₂F₄O₃
MDL Nummer: MFCD00129952
CAS nummer: 652-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d521f4da23743e86b66327294fa20d034cab976&vid=3d521f4da23743e86b66327294fa20d034cab976&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR