Tetrabutylammonium hydroxide 37% in methanol

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T1948-25ML T1948-500ML
TCIAT1948-25MLEA 25.3 EUR
TCIAT1948-25ML TCIAT1948-500ML
Tetrabutylammonium hydroxide 37% in methanol
Tetrabutylammonium hydroxide
Formule: C₁₆H₃₇NO
MW: 259.48 g/mol
Smeltpunt: 29.85
Dichtheid: 0.968
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009425
CAS nummer: 2052-49-5
EINECS: 218-147-6
UN: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e75978a90d30b3709eaf134d63773e77bb784e9&vid=72616a069e7d9e80dbe462d1cbcf5430b4412a43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Concentration (Neutralization titration) 37.0 to 40.7 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR