Triethylene glycol monobenzyl ether ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T1996-25G T1996-5G
TCIAT1996-25GEA 555 EUR
TCIAT1996-25G TCIAT1996-5G
Triethylene glycol monobenzyl ether ≥97.0% (by GC)
Triethylene glycol monobenzyl ether
Formule: C₁₃H₂₀O₄
MDL Nummer: MFCD02093556
CAS nummer: 55489-58-2

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR