4-(Trifluoromethyl)salicylic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T2064-25G T2064-5G
TCIAT2064-25GEA 113 EUR
TCIAT2064-25G TCIAT2064-5G
4-(Trifluoromethyl)salicylic acid ≥98.0% (by GC, titration analysis)
4-(Trifluoromethyl)salicylic acid
Formule: C₈H₅F₃O₃
MDL Nummer: MFCD01529657
CAS nummer: 328-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6b01a7d69f4e476693964b1d099835de5faa40c&vid=c6b01a7d69f4e476693964b1d099835de5faa40c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 176.0 to 181.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR