2-Thiophenesulphonyl chloride ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T2122-25G T2122-5G
TCIAT2122-25GEA 312 EUR
TCIAT2122-25G TCIAT2122-5G
2-Thiophenesulphonyl chloride ≥98.0% (by GC, titration analysis)
2-Thiophenesulphonyl chloride
Formule: C₄H₃ClO₂S₂
Kookpunt: 93°C
Smeltpunt: 33°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00005426
CAS nummer: 16629-19-9
EINECS: 240-677-1
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef2b7a30a3b22b34ab0fdc002caa63c1df4aee60&vid=ef2b7a30a3b22b34ab0fdc002caa63c1df4aee60&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR