2-(Trifluoromethyl)phenylacetonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T2366-25G T2366-5G
TCIAT2366-25GEA 121 EUR
TCIAT2366-25G TCIAT2366-5G
2-(Trifluoromethyl)phenylacetonitrile ≥98.0% (by GC)
2-(Trifluoromethyl)phenylacetonitrile
Formule: C₉H₆F₃N
Kookpunt: 92°C
Smeltpunt: 29…30 °C
MDL Nummer: MFCD00001903
CAS nummer: 3038-47-9
EINECS: 221-239-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80012c18a2d06d799ec0ca0cf7579f0fc3891fd2&vid=80012c18a2d06d799ec0ca0cf7579f0fc3891fd2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR