cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T2423-1G T2423-5G
TCIAT2423-1GEA 58.3 EUR
TCIAT2423-1G TCIAT2423-5G
cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid
Formule: C₈H₁₁F₃O₂
MDL Nummer: MFCD09037361
CAS nummer: 1202578-27-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR