5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-α-cyclodextrin-6-yloxy)phenyl]-21H,23H-porphine

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T2452-10MG
TCIAT2452-10MGEA 234 EUR
TCIAT2452-10MG
5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-α-cyclodextrin-6-yloxy)phenyl]-21H,23H-porphine
5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-alpha-cyclodextrin-6-yloxy)phenyl]-21H,23H-porphine
Formule: C₄₆₈H₇₆₆N₄O₂₄₀
Bewaar temperatuur: Koelkast
CAS nummer: 117194-77-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a72a36df9ce74fb1fd296ea228baa45f81005a74&vid=9d44c9a06c7705b9d9b3169c169bb782f2be15f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Purple to Dark purple to Dark red powder to crystal


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR