Trimethoxy(p-tolyl)silane ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T2560-5G
TCIAT2560-5GEA 108 EUR
TCIAT2560-5G
Trimethoxy(p-tolyl)silane ≥97.0% (by GC)
Trimethoxy(p-tolyl)silane
Formule: C₁₀H₁₆O₃Si
Kookpunt: 143°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00048020
CAS nummer: 17873-01-7

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR