Tricyclohexylphosphonium tetrafluoroborate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

T2585-1G T2585-5G
TCIAT2585-1GEA 20.9 EUR
TCIAT2585-1G TCIAT2585-5G
Tricyclohexylphosphonium tetrafluoroborate ≥98.0% (by titrimetric analysis)
Tricyclohexylphosphonium tetrafluoroborate
Formule: C₁₈H₃₄BF₄P
MW: 368.24 g/mol
MDL Nummer: MFCD03840580
CAS nummer: 58656-04-5
UN: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ab49709982e6b2c5721df45e863a5ce165b378f&vid=2ab49709982e6b2c5721df45e863a5ce165b378f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %
Melting point 168.0 to 172.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR