Tetrakis[N-tetrachlorophthaloyl-(R)-tert-leucinato]dirhodium bis(ethyl acetate) adduct ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T2658-50MG
TCIAT2658-50MGEA 194 EUR
TCIAT2658-50MG
Tetrakis[N-tetrachlorophthaloyl-(R)-tert-leucinato]dirhodium bis(ethyl acetate) adduct ≥98.0% (by HPLC)
Tetrakis[N-tetrachlorophthaloyl-(R)-tert-leucinato]dirhodium bis(ethyl acetate)
Formule: C₆₄H₅₆Cl₁₆N₄O₂₀Rh₂
CAS nummer: 2001054-66-4

Order Now

Specification Test Results

Appearance Yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -315 to -360 deg (C=0.05, Benzene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR