Tris[1-phenylisoquinoline-C2,N]iridium(III) ≥99.0% (by HPLC), purified by sublimation

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Ir(piq)3

T2685-100MG
TCIAT2685-100MGEA 483 EUR
TCIAT2685-100MG
Tris[1-phenylisoquinoline-C2,N]iridium(III) ≥99.0% (by HPLC), purified by sublimation
Tris[1-phenylisoquinoline-C2,N]iridium(III)
Formule: C₄₅H₃₀IrN₃
MW: 804.97 g/mol
MDL Nummer: MFCD09842770
CAS nummer: 435293-93-9

Order Now

Specification Test Results

Appearance Yellow to Amber to Dark red powder to crystal
Purity (HPLC) min. 99.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR