2-(Trifluoromethyl)pyridine-5-carboxaldehyde ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

T2838-1G T2838-5G
TCIAT2838-1GEA 106 EUR
TCIAT2838-1G TCIAT2838-5G
2-(Trifluoromethyl)pyridine-5-carboxaldehyde ≥98.0% (by GC)
2-(Trifluoromethyl)pyridine-5-carboxaldehyde
Formule: C₇H₄F₃NO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01862647
CAS nummer: 386704-12-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afe586afad53d142e34cbd8b9a9e6e931db7e9b7&vid=afe586afad53d142e34cbd8b9a9e6e931db7e9b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 56.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR