N-(Triphenylmethyl)-L-serine methyl ester ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T2870-5G T2870-25G
TCIAT2870-5GEA 41.8 EUR
TCIAT2870-5G TCIAT2870-25G
N-(Triphenylmethyl)-L-serine methyl ester ≥98.0% (by GC, titration analysis)
N-(Triphenylmethyl)-L-serine methyl ester
Formule: C₂₃H₂₃NO₃
MDL Nummer: MFCD00192570
CAS nummer: 4465-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e38da92c3c2ced5514fa459d0a73b6cba95a9cc0&vid=e38da92c3c2ced5514fa459d0a73b6cba95a9cc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 146.0 to 150.0 deg-C
Specific rotation [a]20/D +31.0 to +35.0 deg (C=1、MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR