Trimethyl(pentafluorophenyl)silane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Trimethyl(perfluorophenyl)silane

T3012-1G T3012-5G
TCIAT3012-1GEA 30.8 EUR
TCIAT3012-1G TCIAT3012-5G
Trimethyl(pentafluorophenyl)silane ≥98.0% (by GC)
Trimethyl(pentafluorophenyl)silane
Formule: C₉H₉F₅Si
MW: 240.25 g/mol
Kookpunt: 165°C
MDL Nummer: MFCD00092630
CAS nummer: 1206-46-8
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c658a858b5c2c8211fe564c1c1966cf5d42dea09&vid=c658a858b5c2c8211fe564c1c1966cf5d42dea09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR