Tetrakis(acetonitrile)palladium(II) bis(trifluoromethanesulphonate) ≥95.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Tetrakis(acetonitrile)palladium(II) Ditriflate

T3023-1G
TCIAT3023-1GEA 137 EUR
TCIAT3023-1G
Tetrakis(acetonitrile)palladium(II) bis(trifluoromethanesulphonate) ≥95.0% (by total nitrogen and titration analysis)
Tetrakis(acetonitrile)palladium(II) bis(trifluoromethanesulphonate)
Formule: C₁₀H₁₂F₆N₄O₆PdS₂
MW: 568.76 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD28386116
CAS nummer: 68569-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cad56603c92488cc31bbeff4f1dadb8a736b985f&vid=cad56603c92488cc31bbeff4f1dadb8a736b985f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (Chelometric Titration) min. 95.0 %
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR