N,N,N',N'-Tetraphenyl-1,4-phenylenediamine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T3309-1G T3309-5G
TCIAT3309-1GEA 35.2 EUR
TCIAT3309-1G TCIAT3309-5G
N,N,N',N'-Tetraphenyl-1,4-phenylenediamine ≥98.0% (by GC)
N,N,N',N'-Tetraphenyl-1,4-phenylenediamine
Formule: C₃₀H₂₄N₂
MDL Nummer: MFCD08276846
CAS nummer: 14118-16-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b26743c58ae82cc63f0c64a618eb81392ff5be40&vid=b26743c58ae82cc63f0c64a618eb81392ff5be40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 201.0 to 205.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR